Learn More
CERILLIANT™ Retigabine-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco R0191ML
Catalog No. 01-001-925
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
An internal standard and Certified Spiking Solution™ suitable for quantitation of retigabine levels in urine or other matrices by LC/MS or GC/MS for testing applications in clinical toxicology, forensic analysis, urine drug testing, medication monitoring, or pharmaceutical research. Retigabine, also known as ezogabine, is a new antiepileptic drug developed for treatment of partial seizures. The drug is marketed under the trade names Potiga™ in the US and Trobalt in the EU.
Specifications
Specifications
75-05-8
UN 1648 3 / PGII
307.36
-20°C
6°C (42.8°F) - closed cup (solvent)
C16H14D4FN3O2
100 μg/mL in acetonitrile
Ampule
1 mL
Safety and Handling
Safety and Handling
H332-H319
H312
EINECSNumber : 200-835-2
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation