Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Retinyl palmitate Solution, 100 μg/mL (Ethanol with 0.1% (w/v) BHT), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco V0121ML
Description
Retinyl palmitate is a synthetic alternative for retinyl acetate in vitamin A dietary supplements. Retinyl palmitate is used for treatment of vitamin A defficiency under brand names such as Aquasol A™ and Palmitate A and is a constituent of some skin care products.Specifications
64-17-5, 79-81-2 | |
MFCD00019414 | |
100 μg/mL (ethanol with 0.1% (w/v) BHT) | |
524.86 | |
-70°C | |
14°C (57.2°F) |
C36H60O2 | |
UN 1170 - class 3 - PG 2 - Ethanol, solution | |
Retinyl palmitate solution | |
Ampule | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation