Learn More
CERILLIANT™ Salicylic Acid Impurity Mixture Solution, 10-500 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco S1001ML
Catalog No. 01-001-996
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Salicylic Acid belongs to a group of phenols found in plants that are characterized by having an aromatic ring with a hydroxyl group or its derivative. Rice, crabgrass, green foxtail, barley, and soybean are some agriculturally important species of plants that show the presence of salicylic acid. It is proposed to act as a signal transducer or messenger during conditions of environmental stress in plants, thereby aiding the plant in acquiring multiple stress tolerance. It is also used in combination with benzoyl peroxide in the treatment of acne vulgaris.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
127.12
-20°C
20°C (68°F)
C6H7O3
10 to 500 μg/mL (MeOH:water:acetic acid (70:30:4))
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation