Learn More
CERILLIANT™ SDB-005 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco S1191ML
Catalog No. 01-002-013
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
SDB-005 is a synthetic cannabinoid that has emerged on the illicit designer drug market. This certified Snap-N-Spike™ solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications.For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.
Specifications
Specifications
75-05-8
MFCD30718277
100 μg/mL in acetonitrile
Ampule
1 mL
C23H22N2O2
UN 1648 3 / PGII
358.43
-20°C
2°C (35.6°F) - closed cup
Safety and Handling
Safety and Handling
H332-H319
H312
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation