Learn More
CERILLIANT™ Tapentadol-D3-beta-D-glucuronide Solution, 100 μg/mL in Acetonitrile:Water, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T0671ML
Description
Tapentadol-β-D-glucuronide is the primary urinary metabolite of tapentadol, the first new drug of the centrally-acting analgesic class approved in the US for pain management in more than 25 years. This new internal standard is suitable for many LC/MS or GC/MS applications including pain prescription monitoring, forensic testing, clinical toxicology, urine drug testing or isotope dilution methods.Specifications
75-05-8 | |
UN 1648 3 / PGII | |
400.48 | |
-20°C | |
13°C (55.4°F) - closed cup |
C20D3H28NO7 | |
100 μg/mL in acetonitrile:water | |
Ampule | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation