Learn More
CERILLIANT™ Testosterone CRM in Serum Solution, 2000 ng/dL (Stripped Serum), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T5101ML
Description
Certified reference material of testosterone in stripped serum for clinical research applications by mass spectrometry. Individual levels of testosterone in stripped serum are offered separately at concentrations ranging from 2 to 2,000 ng/dL. Each individual level is ready to use as a linearity standard or for calibration verification in LC-MS/MS testosterone methods. Separate options for a blank and 13C- or deuterium labeled internal standards of testosterone are also available.Specifications
C19H28O2 | |
288.42 | |
-20°C |
2000 ng/dL (stripped serum) | |
Ampule | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation