Learn More
CERILLIANT™ Tetrahydrofuranyl fentanyl-13C6 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco T1310.5ML
Catalog No. 01-002-098
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This stable-labeled internal standard is suitable for quantitation of tetrahydrofuranyl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, or isotope dilution methods. Tetrahydrofuranyl fentanyl is an opioid analgesic analog of fentanyl and has been sold as a designer drug on the illicit drug market.
Specifications
Specifications
67-56-1
100 μg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
C1813C6H30N2O2 · HCl
420.92
0.5 mL
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation