Learn More
CERILLIANT™ TH-PVP Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco T1211ML
Catalog No. 01-002-091
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
TH-PVP is a stimulant drug of the cathinone class and has emerged on the illicit drug market as a designer drug. This Certified Spiking Solution is suitable as starting material in calibrators and controls for GC/MS or LC/MS testing applications in forensic analysis, urine drug testing or clinical toxicology.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
321.88
-20°C
C19H27NO · HCl
1 mg/mL in methanol (as free base)
Ampule
1 mL
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation