Learn More
CERILLIANT™ Theophylline Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco IMPC051011ML
Description
A Certified Spiking Solution™ suitable for use as starting material in the preparation of calibrators and controls for theophylline testing methods by LC-MS/MS or GC-MS. Theophylline, also known as dimethylxanthine, is a naturally occurring stimulant of the xanthine family. Sold under the trade name Theolair, theophylline is used for treatment of asthma and other respiratory diseases.Specifications
67-56-1, 58-55-9 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
180.16 | |
2 to 8°C | |
9.7°C (49.5°F) - closed cup |
C7H8N4O2 | |
1.0 mg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation