Learn More
CERILLIANT™ Thiamine Impurity Mixture Solution, 100 μg/mL (Methanol:1% Acetic Acid 50:50, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T1341ML
Specifications
67-56-1, 64-19-7 | |
100 μg/mL (methanol:1% Acetic Acid 50:50 | |
Ampule | |
1 mL |
C11H15ClN4OS · HCl | |
323.24 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H314-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation