Learn More
CERILLIANT™ Thiamine pyrophosphate (Vitamin B1 pyrophosphate) Solution, 1 mg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco V0531ML
Description
Thiamine pyrophosphate, also known as TPP or ThPP, is the biologically active form of vitamin B1. It is usually measured in whole blood or in plasma. While plasma thiamine concentration reflects recent intake rather than body stores, whole blood is the preferred specimen for thiamine assessment. This Certified Spiking Solution™ is suitable for use as a starting material in preparation of linearity standards, calibrators, and controls in a variety of testing applications from diagnostic testing of thiamine defficiency to quality control testing of thiamine-based dietary supplements, fortified foods or pharmaceutical preparations.Specifications
67-56-1, 154-87-0 | |
UN 1992 6.1(3) / PGIII | |
460.77 | |
-20°C | |
24°C (75.2°F) |
C12H19ClN4O7P2S | |
1 mg/mL (50:50 methanol:water) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation