Learn More
CERILLIANT™ (+/-)-threo-3,4-Dichloromethylphenidate HCl, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1401ML
Description
3,4-Dichloromethylphenidate, also known as 3,4-CTMP, is a structural analogue of methylphenidate (Ritalin). 3,4-CTMP is reportedly seven times more potent than methylphenidate and is sold online as a novel psychoactive stimulant. This certified Snap-N-Spike™ solution standard is applicable for use in LC/MS or GC/MS applications such as urine drug testing, clinical toxicology and forensic analysis.Specifications
67-56-1, 214149-42-5 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
338.66 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C14H17Cl2NO2 · HCl | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation