Learn More
CERILLIANT™ Tiagabine-D6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco T0821ML
Catalog No. 01-002-079
Description
Description
A Certified Spiking Solution™ suitable for use as an internal standard in LC/MS or GC/MS tiagabine testing methods for clinical toxicology, forensic analysis, urine drug testing, medication monitoring, or pharmaceutical research applications. Tiagabine, marketed as the antiepileptic drug Gabitril™, is used in the treatment of anxiety disorders and neuropathic path including fibromyalgia. The drug has also been approved by US FDA as an adjunctive treatment for partial seizures in ages 12 and up.
Specifications
Specifications
67-56-1, 1217808-68-8
UN 1230 - class 3 - PG 2 - Methanol, solution
418.05
-20°C
9.7°C (49.5°F)
C20H19D6NO2S2 · HCl
100 μg/mL in methanol (as free base)
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation