Learn More
CERILLIANT™ Trifluoperazine N-ß-D-glucuronide Solution, 100 μg/mL in Methanol:Water (1:1), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T1181ML
Description
Trifluoperazine N-β-D-glucuronide is a urinary and blood metabolite of trifluoperazine, a typical antipsychotic used for treating schizophrenia. Trifluoperazine is also indicated for use in patients with agitation, severe anxiety, or behavioral problems. This Snap-N-Spike™; reference material is suitable for use as a hydrolysis control in mass spectrometry-based testing applications of trifluoperazine in clinical toxicology, forensic analysis, therapeutic drug monitoring, and urine drug testing.Specifications
67-56-1, 165602-90-4 | |
UN 1986 6.1(3) / PGIII | |
583.62 | |
-70°C | |
24°C (75.2°F) |
C27H32F3N3O6S | |
100 μg/mL in methanol:water (1:1) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation