Learn More
CERILLIANT™ Triflupromazine-D6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T1291ML
Description
A stable-labeled internal standard suitable for quantitation of Triflupromazine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, isotope dilution methods, or therapeutic drug monitoring. Triflupromazine, sold as Vesprin, is an antipsychotic medication of the phenothiazine class.Specifications
67-56-1, 1416711-36-8 | |
100 μg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C18H13D6F3N2S · HCl | |
394.91 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation