Learn More
CERILLIANT™ Triflupromazine Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T1281ML
Description
Triflupromazine, sold as Vesprin, is an antipsychotic medication with high potential for side effects. This Certified Spiking Solution™ is suitable for LC/MS or GC/MS methods in triflupromazine testing for therapeutic drug monitoring, forensic analysis, clinical toxicology, or urine drug testing.Specifications
67-56-1, 1098-60-8 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
388.88 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C18H19F3N2S · HCl | |
1 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation