Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Trimipramine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T9041ML
Description
Trimipramine is a tricyclic antidepressant prescribed for treatment of depression, insomnia, and chronic pain or as an adjunctive therapy for alcohol or opioid withdrawal. This certified solution standard is applicable for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC-MS/MS or GC/MS. Trimipramine is marketed under the trade names Surmontil™, Rhotrimine, and Stangyl.Specifications
67-56-1, 739-71-9 | |
MFCD00242595 | |
1.0 mg/mL in methanol | |
Ampule | |
1 mL |
C20H26N2 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
294.43 | |
2 to 8°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation