Learn More
CERILLIANT™ U-48800-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco U0121ML
Description
A certified Snap-N-Shoot™ solution suitable for quantification of U-48800 by LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. U-48800 is a highly potent opioid agonist, not structurally related to fentanyl, that has been confirmed in opioid overdoses and deaths. Novel synthetic opioids (NSOs) created to circumvent drug control laws such as U-48800 have no recognized therapeutic use, are clandestinely manufactured and sold on conventional or dark web.Specifications
67-56-1 | |
100 μg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C17H21D3Cl2N2O · HCl | |
382.77 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation