Learn More
CERILLIANT™ U-49900 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco U0091ML
Description
U-49900 is a highly potent opioid analgesic that has been confirmed in many opioid overdoses. This Certified Spiking Solution™ is suitable as starting material for use in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology, urine drug testing or forensic analysis applications.This Cerilliant product is part of the Centers for Disease Control and Prevention (CDC) Opioid CRM Kit including more than 40 CRMs to be used as part of CDC's laboratory response to the opioid epidemic.Specifications
67-56-1, 67579-76-4 | |
1.0 mg/mL in methanol | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C18H26Cl2N2O | |
357.32 | |
1 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation