Learn More
CERILLIANT™ U-50488 mesylate Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco U0071ML
Description
U-50488 is a potent opioid agonist, not structurally related to fentanyl. U-50488 belongs to the class of novel synthetic opioids (NSOs) created to circumvent drug control laws with no recognized therapeutic use. NSOs are clandestinely manufactured and sold on conventional or dark web. This Certified Spiking Solution™ is offered as DEA-exempt solution standard and applicable for use in urine drug testing, clinical toxicology, or forensic analysis applications by LC-MS/MS or GC/MS.Specifications
67-56-1, 83913-06-8 | |
1 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C19H26Cl2N2O · CH3SO3H | |
465.43 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation