Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Ursodeoxycholic acid-D4, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco U0021ML
Description
A stable-labeled internal standard suitable for quantitation ursodeoxycholic acid (UDCA) levels by LC/MS or GC/MS for clinical diagnostic testing applications such as screening for inborn errors of metabolism or monitoring bile acid therapy. Ursodeoxycholic acid (UDCA) is a secondary bile acid that helps regulate cholesterol.Specifications
67-56-1, 347841-46-7 | |
MFCD01074179 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C24H36D4O4 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
396.60 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation