Learn More
CERILLIANT™ W-18-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco W0051ML
Description
W-18 is a fentanyl-related designer drug that first appeared on the illicit drug market in the 2010s. This stable-labeled internal Snap-N-Shoot™ solution standard is suitable for quantitation of W-18 levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, or isotope dilution methods.Specifications
75-05-8 | |
100 μg/mL in acetonitrile | |
Ampule | |
1 mL |
C19H16D4ClN3O4S | |
425.92 | |
-20°C | |
2°C (35.6°F) - closed cup |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation