Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ W-18 Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco W0041ML
Description
A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for W-18 LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. W-18 is a fentanyl-related designer drug that first appeared on the illicit drug market in the 2010s.Specifications
75-05-8, 93101-02-1 | |
MFCD24369672 | |
1 mg/mL in acetonitrile | |
Ampule | |
1 mL |
C19H20ClN3O4S | |
UN 1648 3 / PGII | |
421.90 | |
-20°C | |
2°C (35.6°F) - closed cup |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation