Learn More
CERILLIANT™ Warfarin Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco W0031ML
Description
Warfarin is an anticoagulant sold under the trade names Coumadin™ and Jantoven™. Warfarin is also used a pesticide, most often as rat poison. This Snap-N-Spike™ reference solution is applicable for use in clinical toxicology, forensic analysis, therapeutic drug monitoring or environmental monitoring and diagnostic testing applications by LC-MS/MS or GC/MSSpecifications
75-05-8, 81-81-2 | |
UN 1648 3 / PGII | |
308.33 | |
2 to 8°C | |
2°C (35.6°F) - closed cup |
C19H16O4 | |
1.0 mg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H312-H319-H331-H360D-H373
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation