Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Zidovudine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco Z0111ML
Description
Zidovudine, also known as azidothymidine (AZT), is a nucleoside analog reverse-transcriptase inhibitor (NRTI) antiretroviral used for the treatment of human immunodefficiency virus (HIV) infection. Therapeutic drug monitoring (TDM) of zidovudine may be requested to evaluate its toxicity, assess virological failure, guide dosing regimens, or investigate drug-drug interactions. This Certified Spiking Solution™ is suitable for use as starting material in the preparation of calibrators, controls, or linearity standards for a variety of LC-MS/MS or GC/MS zidovudine testing applications including TDM and clinical and diagnostic testing.Specifications
67-56-1, 30516-87-1 | |
MFCD25542524 | |
1.0 mg/mL in methanol | |
Ampule | |
1 mL |
C10H13N5O4 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
267.24 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation