Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Ziprasidone Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco Z0181ML
Description
A Certified Spiking Solution™; applicable to use in forensic drug analysis or calibrator preparation by LC/MS or GC/MS. Ziprasidone is sold under the trade names Geodon™ and Zeldox for treatment of schizophrenia and bipolar disorder. Recent studies have suggested that Ziprasidone may increase the risk of Type II diabetes and hyperglycemia.Specifications
67-56-1, 122883-93-6 | |
MFCD06795476 | |
1.0 mg/mL in methanol | |
Ampule | |
1 mL |
C21H21ClN4OS · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
449.40 | |
-70°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation