Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cerium foil, 0.62mm (0.024 in.) thick, 99.9% (REO)

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7440-45-1
Ce
140.116
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium
23974
CHEBI:33369
cerium
[Ce]
7440-45-1
Ce
140.116
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium
23974
CHEBI:33369
cerium
[Ce]
  • Packaged under argon
Quantity 50 x 50 mm
Weight ≈2.6g/25x25mm
UN Number UN1333
Odor Odorless
Packaging Mylar under Argon
Physical Form Foil
Percent Purity 99.9%
Color Silver to Gray
Chemical Name or Material Cerium foil, 0.62mm (0.024 in.) thick
Assay (REO)

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

EINECSNumber : 231-154-9

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures