Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium ICP Standard, 1mL = 1mg Ce (1,000ppm Ce), Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
10294-41-4,7697-37-2,7732-18-5
CeH12N3O15
434.218
QQZMWMKOWKGPQY-UHFFFAOYSA-N
16211466
cerium(3+);trinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]
10294-41-4,7697-37-2,7732-18-5
CeH12N3O15
434.218
QQZMWMKOWKGPQY-UHFFFAOYSA-N
16211466
cerium(3+);trinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]
CAS Min % 93.0,4.21,0.29
CAS Max % 97.0,4.39,0.3
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 1,000ppm Ce
Equivalence 1 mL = 1 mg Ce
Color Colorless
Quantity 100mL
Chemical Name or Material Cerium ICP Standard