Alfa Aesar™ Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO)

Manufacturer: Alfa Aesar™ 1132409

Quantity 10g

 View more versions of this product

Catalog No. AA1132409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 537-00-8
    Formula Weight 344.28 (317.26 Anhydrous)
    MDL Number MFCD00051060
    Chemical Name or Material Cerium(III) acetate sesquihydrate
    Grade Reacton™

    Specifications

    Formula Weight 344.28 (317.26 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline Aggregates
    Health Hazard 2 GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.
    TSCA TSCA
    MDL Number MFCD00051060
    Melting Point 308°C (dec.)
    Health Hazard 1 Danger
    CAS 537-00-8
    Quantity 10g
    Chemical Name or Material Cerium(III) acetate sesquihydrate
    Assay 99.99%, (REO)
    Grade Reacton™

    GHS H Statement
    H318-H335-H315
    Causes serious eye damage.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit