Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) ammonium nitrate tetrahydrate, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
13083-04-0
CeH16N7O19
558.274
MFCD00210608
AIVZRMAXALRCHB-UHFFFAOYSA-N
ce.3no3.2hno3.4h2o.2nh3,ammonium cerium iii nitrate tetrahydrate,cerous ammonium nitrate, hydrated, reagent,cerium iii ammonium nitrate tetrahydrate, reagent grade,cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
91886599
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate
N.N.[N+](=O)(O)[O-].[N+](=O)(O)[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ce+3]
13083-04-0
CeH16N7O19
558.274
MFCD00210608
AIVZRMAXALRCHB-UHFFFAOYSA-N
ce.3no3.2hno3.4h2o.2nh3,ammonium cerium iii nitrate tetrahydrate,cerous ammonium nitrate, hydrated, reagent,cerium iii ammonium nitrate tetrahydrate, reagent grade,cerium 3+ bis nitric acid diamine tetrahydrate trinitrate
91886599
azane;cerium(3+);nitric acid;trinitrate;tetrahydrate
N.N.[N+](=O)(O)[O-].[N+](=O)(O)[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ce+3]
Appearance White crystalline
Solubility Information Soluble in water.
Formula Weight 558.28 (486.22 Anhydrous)
Physical Form Crystalline
Grade Reagent
Color White
Quantity 100g
Chemical Name or Material Cerium(III) ammonium nitrate tetrahydrate, For reagent grade

P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H272-H315-H319

DOTInformation : DOT Class: 5.1, Packaging Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures