Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) chloride, 99.9%, pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
7790-86-5
CeCl3
246.48
MFCD00010929
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium iii chloride,cerous chloride,cerium iii chloride, anhydrous,cerium-144 chloride,cerium chloride generic,cerium-chloride,cerium chloride octahydrate,cerous iii-chloride
24636
CHEBI:35458
cerium(3+);trichloride
[Cl-].[Cl-].[Cl-].[Ce+3]
7790-86-5
CeCl3
246.48
MFCD00010929
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium iii chloride,cerous chloride,cerium iii chloride, anhydrous,cerium-144 chloride,cerium chloride generic,cerium-chloride,cerium chloride octahydrate,cerous iii-chloride
24636
CHEBI:35458
cerium(3+);trichloride
[Cl-].[Cl-].[Cl-].[Ce+3]
Assay Percent Range 99.90%
Linear Formula CeCl3
Merck Index 15, 1996
Solubility Information Solubility in water: soluble
Formula Weight 246.48
Physical Form Powder
Percent Purity 99.9%
Grade Pure
Packaging Glass bottle
Solubility Solubility in water: soluble.
Boiling Point 1727.0°C
Color White to Gray
Melting Point 848.0°C
Quantity 100g
Chemical Name or Material Cerium(III) chloride, Anhdrous
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 232-227-8