Alfa Aesar™ Cerium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1366218

Quantity 50g

 View more versions of this product

Catalog No. AA1366218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7758-88-5
    Formula Weight 197.12
    MDL Number MFCD00010932
    Merck Index 14,1998
    Chemical Name or Material Cerium(III) fluoride, Anhydrous
    Grade Reacton™

    Specifications

    Physical Form Lumps
    Formula Weight 197.12
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7758-88-5
    Boiling Point ≈2300°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number FK6125000
    Sensitivity Hygroscopic
    MDL Number MFCD00010932
    Merck Index 14,1998
    Density 6.16g/mL
    TSCA TSCA
    Odor Odorless
    Quantity 50g
    Melting Point 1460°C
    Health Hazard 1 Warning
    Chemical Name or Material Cerium(III) fluoride, Anhydrous
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit