Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) iodide, ultra dry, 99.9% (REO), Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
7790-87-6
CeI3
MFCD00049423
Cerium triiodide
7790-87-6
CeI3
MFCD00049423
Cerium triiodide

Used as a pharmaceutical intermediate.

Sensitivity Hygroscopic; Light sensitive
Quantity 25g
Melting Point 766°C
Packaging Ampouled under argon
Solubility Information Soluble in water
Formula Weight 520.83
Physical Form Powder
Percent Purity 99.9%
Chemical Name or Material Cerium(III) iodide

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H360

EINECSNumber : 232-228-3

TSCA : No

Recommended Storage : Ambient temperatures