Cerium(III) nitrate hexahydrate, 99.99% (REO) , Alfa Aesar™, REacton™

Manufacturer: Alfa Aesar 1133030

Quantity 250g

 View more versions of this product

Catalog No. AA1133030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10294-41-4
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.99% (REO)
    Molecular Formula CeN3O9.6H2O N5Na2O
    Formula Weight 434.23 (326.13 Anhydrous)
    MDL Number MFCD00149631
    Chemical Name or Material Cerium(III) nitrate hexahydrate

    Specifications

    Formula Weight 434.23 (326.13 Anhydrous)
    Quantity 250g
    Storage Note 1 Hygroscopic
    EINECS Number 233-297-2
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.99% (REO)
    Molecular Formula CeN3O9.6H2O N5Na2O
    MDL Number MFCD00149631
    CAS 10294-41-4
    Chemical Name or Material Cerium(III) nitrate hexahydrate, hexahydrate

    Hygroscopic

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit