Learn More
Alfa Aesar™ Cerium(III) perchlorate hexahydrate, Reagent Grade
Manufacturer:  Alfa Aesar™ 1165418
Quantity 50g
 View more versions of this product
Catalog No. AA1165418
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
14017-47-1
MFCD00150205
Cerium(III) perchlorate hexahydrate
546.56 (438.47 Anhydrous)
UN1481
Reagent Grade
Specifications
14017-47-1
546.56 (438.47 Anhydrous)
UN1481
Moisture Sensitive
Cerium(III) perchlorate hexahydrate
Crystalline
MFCD00150205
Odorless
50g
Reagent Grade
Safety and Handling
SDS & Safety Documents
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
TSCA : TSCA