Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) sulfate, 99.9%, (REO=Rare Earth Oxides), Acros Organics

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
13454-94-9
Ce2O12S3
568.43
MFCD00010931
OZECDDHOAMNMQI-UHFFFAOYSA-H
cerium sulfate,cerous sulfate,cerium iii sulfate,cerium sulphate,dicerium trisulfate,cerium sesquisulfate,unii-0vf70o14r1,sulfuric acid, cerium salt,dicerium trisulphate,sulphuric acid, cerium salt
159674
cerium(3+);trisulfate
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+3].[Ce+3]
13454-94-9
Ce2O12S3
568.43
MFCD00010931
OZECDDHOAMNMQI-UHFFFAOYSA-H
cerium sulfate,cerous sulfate,cerium iii sulfate,cerium sulphate,dicerium trisulfate,cerium sesquisulfate,unii-0vf70o14r1,sulfuric acid, cerium salt,dicerium trisulphate,sulphuric acid, cerium salt
159674
cerium(3+);trisulfate
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+3].[Ce+3]
For ultra trace analysis
Assay Percent Range 46.0 to 50% (Ce) (Complexometry)
Linear Formula Ce2(SO4)3
Merck Index 15, 2001
Solubility Information Solubility in water: slightly soluble
Formula Weight 568.43
Physical Form Liquid, May Contain Some Sediment
Percent Purity 99.90%
Grade (REO=Rare Earth Oxides)
Packaging Glass bottle
Total Trace Metal Impurities 0.01% max.
Color Colorless
Quantity 250g
Chemical Name or Material Cerium(III) sulfate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-644-6