Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cerium iron oxide, 20% in H2O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted

Manufacturer:  Alfa Aesar™ 4728336

Quantity 500g

 View more versions of this product

Catalog No. AA4728336


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD02687051
Cerium and iron mixed metal oxide in water at low pH
Cerium iron oxide, 20% in H2O, nanoparticle dispersion

Specifications

Liquid
Cerium and iron mixed metal oxide in water at low pH
1.23g/mL
<5.0nm APS Number Weighted
Low
Cerium iron oxide, 20% in H2O, nanoparticle dispersion
MFCD02687051
CeO2:Fe2O3; =50:50 wt%
Petroleum-like
Red-brown
500g

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H335
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P304+P340-P362-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit