Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol
Click to view available options
Quantity:
5g
25g
100g
Specifications
Concentration or Composition (by Analyte or Components) | 18 to 20% w/w in 2-methoxyethanol |
Sensitivity | Moisture sensitive |
Density | 1.02 g/mL |
Quantity | 25g |
Color | Yellow |
Linear Formula | Ce[OCH2CH2OCH3]4 |
UN Number | UN1188 |
Solubility Information | Reacts with air. |
Formula Weight | 440.47 |
Physical Form | Liquid |
Show More |
Safety and Handling
GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312+H332-H314-H335-H360FD-H370-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
TSCA : No
Recommended Storage : Ambient temperatures