Learn More
Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals
Description
Cerium(IV) ammonium nitrate is used to offers a novel route to tartronic acid derivatives via facile oxygenation of malonates. It is also used as for oxidative addition reactions of electrophilic radicals to alkenes, enabling intermolecular and intramolecular carbon-carbon and carbon-heteroatom bond formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow to Orange |
Physical Form | Crystalline |
Quantity | 10 g |
Assay Percent Range | 99.5% (REO) |
Solubility Information | Soluble in water. |
Formula Weight | 548.23 |
Odor | Pungent |
Chemical Name or Material | Cerium(IV) ammonium nitrate |
Safety and Handling
Hazard Category | H272-H290-H302-H314-H317-H335 |
Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: II |
EINECSNumber | 240-827-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.