Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min, Alfa Aesar™

Manufacturer: Alfa Aesar 40194A7

Quantity 5kg

 View more versions of this product

Catalog No. AA40194A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10378-47-9
    Physical Form Crystalline
    Assay Percent Range 94% min.
    Molecular Formula (NH4)4Ce(SO4)4·2H2O
    Formula Weight 632.56 (596.53 Anhydrous)
    MDL Number MFCD00149417
    Synonym Ammonium cerium sulfate
    Chemical Name or Material Cerium(IV) ammonium sulfate dihydrate, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 632.56 (596.53 Anhydrous)
    Quantity 5kg
    Color Yellow
    Physical Form Crystalline
    Assay Percent Range 94% min.
    Molecular Formula (NH4)4Ce(SO4)4·2H2O
    Appearance Yellow crystalline
    MDL Number MFCD00149417
    Synonym Ammonium cerium sulfate
    CAS 10378-47-9
    Health Hazard 1 H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Cerium(IV) ammonium sulfate dihydrate, For analysis ACS
    Grade ACS Grade

    H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit