Alfa Aesar™ Cerium(IV) isopropoxide isopropanol adduct, Ce 31.5-32.4%

Manufacturer: Alfa Aesar™ 4565703

Quantity 1g

 View more versions of this product

Catalog No. AA4565703

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C12H28CeO4·C3H8O
    Linear Formula Ce[OCH(CH3)2]4·C3H7OH
    Formula Weight 436.57
    MDL Number MFCD12547727
    UN Number UN3180
    Chemical Name or Material Cerium (IV) isopropoxide isopropanol adduct

    Specifications

    Formula Weight 436.57
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula Ce[OCH(CH3)2]4·C3H7OH
    UN Number UN3180
    Health Hazard 1 Danger
    Physical Form Powder
    Molecular Formula C12H28CeO4·C3H8O
    MDL Number MFCD12547727
    Health Hazard 2 GHS H Statement
    H228-H314-H318
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, CORROSIVE, INORGANIC, N.O.S.
    Concentration Ce 31.5 to 32.4%
    Sensitivity Moisture Sensitive
    Color Yellow to orange-red
    Quantity 1g
    Chemical Name or Material Cerium (IV) isopropoxide isopropanol adduct

    GHS H Statement
    H228-H314-H318
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit