Cerium(IV) oxide, 18% in H2O, colloidal dispersion with dispersant, NanoTek™, Alfa Aesar™

For Use As a Cerium Oxide Water Borne Dispersion Suitable For Use in Ultrafine Alkaline Side Polishing Slurries

Manufacturer: Alfa Aesar 4491122

Quantity 100g

 View more versions of this product

Catalog No. AA4491122

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1306-38-3
    Physical Form Liquid
    Molecular Formula CeO2
    Formula Weight 172.12
    MDL Number MFCD00010933
    Chemical Name or Material Cerium(IV) oxide, CE-6082, 18% in H2O, colloidal dispersion with dispersant

    Specifications

    Physical Form Liquid
    Formula Weight 172.12
    Color Pale yellow to pale brown
    Quantity 100g
    Molecular Formula CeO2
    MDL Number MFCD00010933
    CAS 1306-38-3
    Health Hazard 1 H303 : May be harmful if swallowed.
    Chemical Name or Material Cerium(IV) oxide, CE-6082, 18% in H2O, colloidal dispersion with dispersant

    H303 : May be harmful if swallowed.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit