Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) oxide, 20% in H2O, colloidal dispersion in water with dispersant, NanoTek™, Alfa Aesar™

For Use As a Cerium Oxide Water Borne Dispersion Suitable For Use in Alkaline Side Polishing Slurries. Submicron Particle Size Makes For Good Removal Rate With Minimal Subsurface Damage

Manufacturer:  Alfa Aesar 4491036

Quantity 500g

 View more versions of this product

Catalog No. AA4491036

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1306-38-3
CeO2
MFCD00010933
Dry Powder
172.12
Cerium(IV) oxide, CE-6080, 20% in H2O, colloidal dispersion with dispersant

Specifications

1306-38-3
CeO2
MFCD00010933
Cerium(IV) oxide, CE-6080, 20% in H2O, colloidal dispersion with dispersant
Dry Powder
172.12
500g

Safety and Handling

H303 : May be harmful if swallowed.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit