Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number AC199121000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
1306-38-3
CeO2
172.12
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
73963
CHEBI:79089
dioxocerium
O=[Ce]=O
1306-38-3
CeO2
172.12
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
73963
CHEBI:79089
dioxocerium
O=[Ce]=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 2600.0°C
Color White to Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9%
Linear Formula CeO2
Fieser 06,99
Merck Index 15,1988
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 172.12
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 7.13
Density 7.1300g/mL
Chemical Name or Material Cerium(IV) oxide, 99.9%
Show More Show Less
EINECSNumber 215-150-4
RTECSNumber FK6310000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit