Alfa Aesar™ Cerium(IV) oxide, doped with 18% zirconium, 13% in H2O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted

Manufacturer: Alfa Aesar™ 4725936

Quantity 500g

 View more versions of this product

Catalog No. AA4725936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1306-38-3
    MDL Number MFCD00010933
    Chemical Name or Material Cerium(IV) oxide, Doped with 18% zirconium, 13% in H2O, nanoparticle dispersion

    Specifications

    pH High
    Odor Odorless
    TSCA TSCA
    Particle Size <5.0nm APS Number Weighted
    Physical Form Dispersion
    MDL Number MFCD00010933
    CAS 1306-38-3
    Color Red-brown
    Quantity 500g
    Chemical Name or Material Cerium(IV) oxide, Doped with 18% zirconium, 13% in H2O, nanoparticle dispersion

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit