Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cerium(IV) oxide pellets, 99.9% (metals basis)

Sintered pellets suitable for vacuum deposition

Manufacturer:  Alfa Aesar™ 4065022

Quantity 100g

 View more versions of this product

Catalog No. AA4065022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1306-38-3
MFCD00010933
Cerium(IV) oxide pellets, approx. 9mm (0.35 in.) dia. x 5mm (0.20 in.) thick
172.12
14,1989

Specifications

1306-38-3
MFCD00010933
Odorless
2600°C
Cerium(IV) oxide pellets, approx. 9mm (0.35 in.) dia. x 5mm (0.20 in.) thick
172.12
14,1989
Pale yellow
100g
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FK6310000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit