Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cerium(IV) and Perchloric acid, Etchant Solution, Ce(IV) concentration 0.22N

Etchant solution for printed circuits and for chrome or nichrome photoresists

Manufacturer:  Alfa Aesar™ 42108K2

Quantity 1L

 View more versions of this product

Catalog No. AA42108K2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD01866051
UN3093
Cerium(IV) and Perchloric acid, Etchant Solution

Specifications

Liquid
UN3093
1L
MFCD01866051
Ce(IV) concentration 0.22N
Cerium(IV) and Perchloric acid, Etchant Solution

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit