Alfa Aesar™ Cerium(IV) and Perchloric acid, Etchant Solution, Ce(IV) concentration 0.22N

Etchant solution for printed circuits and for chrome or nichrome photoresists

Manufacturer: Alfa Aesar™ 42108K7

Quantity 4L

 View more versions of this product

Catalog No. AA42108K7

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01866051
    UN Number UN3093
    Chemical Name or Material Cerium(IV) and Perchloric acid, Etchant Solution

    Specifications

    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD01866051
    UN Number UN3093
    Health Hazard 1 Danger
    TSCA TSCA
    Quantity 4L
    Health Hazard 2 GHS H Statement
    H272-H314-H318
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Concentration Ce(IV) concentration 0.22N
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
    Chemical Name or Material Cerium(IV) and Perchloric acid, Etchant Solution

    GHS H Statement
    H272-H314-H318
    May intensify fire
    oxidizer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit