Alfa Aesar™ Cerium(IV) sulfate hydrate, 98%

Manufacturer: Alfa Aesar™ A1014218

Quantity 50g

 View more versions of this product

Catalog No. AAA1014218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95838-16-7
    Molecular Formula Ce(SO4)2·xH2O
    Formula Weight 332.24 (Anhydrous)
    MDL Number MFCD00149427
    UN Number UN3085
    Merck Index 14,1990
    Synonym Ceric sulfate hydrate
    Chemical Name or Material Cerium (IV) sulfate hydrate

    Specifications

    Formula Weight 332.24 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00149427
    TSCA TSCA
    CAS 95838-16-7
    Merck Index 14,1990
    Quantity 50g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
    Molecular Formula Ce(SO4)2·xH2O
    UN Number UN3085
    Synonym Ceric sulfate hydrate
    Melting Point 195°C (dec.)
    Health Hazard 2 GHS H Statement
    H314-H272
    Causes severe skin burns and eye damage.
    May intensify fire
    oxidizer.
    RTECS Number WS6960000
    Chemical Name or Material Cerium (IV) sulfate hydrate
    Assay 98%

    Description

    Applications
    Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

    Solubility
    Soluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. keep container tightly closed

    GHS H Statement
    H314-H272
    Causes severe skin burns and eye damage.
    May intensify fire
    oxidizer.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit