Alfa Aesar™ Cerium(IV) sulfate hydrate, REacton™, 99% (REO)

Manufacturer: Alfa Aesar™ 1132322

Quantity 100g

 View more versions of this product

Catalog No. AA1132322

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 95838-16-7
    Formula Weight 332.24 (Anhydrous)
    MDL Number MFCD00149427
    UN Number UN3085
    Chemical Name or Material Cerium(IV) sulfate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 332.24 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00149427
    TSCA TSCA
    Physical Form Crystalline
    CAS 95838-16-7
    UN Number UN3085
    Quantity 100g
    Health Hazard 1 Danger
    Density 3.01g/mL
    Sensitivity Moisture Sensitive
    Melting Point 195°C (dec.)
    Health Hazard 2 GHS H Statement
    H314-H272
    Causes severe skin burns and eye damage.
    May intensify fire
    oxidizer.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
    RTECS Number WS6960000
    Chemical Name or Material Cerium(IV) sulfate hydrate
    Assay 99%, (REO)
    Grade Reacton™

    GHS H Statement
    H314-H272
    Causes severe skin burns and eye damage.
    May intensify fire
    oxidizer.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit